Accessories for Compresstome® VF200/VF300 Buffer Tray
Catalog # 77435-446
Supplier: PRECISIONARY INSTRUMENTS LLC
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Compresstome® VF-200/VF-300 buffer tray
- Cat. No.:77435-446
Specifications
About this item
This buffer tray is compatable with legacy model VF-200 and VF-300 Compresstome® models.
- Buffer tray made of high-quality, durable Lexan™ polycarbonate
- Full one year warranty
Packaging: Set includes buffer tray, one set of screws, and one gasket.