Specifications
- Description:Rotary microtome RF-1000 side panel
- Cat. No.:77435-310
Specifications
About this item
A replacement part for the external control panel on the RF-1000 rotary microtome.
- Includes side control panel and cable
Control panel allows for control of slice thickness adjustments, movement of the specimen head, and start/stop of automated cutting.