Order Entry
Puerto Rico
ContactUsLinkComponent
Rotary Microtome Low Profile Blades
Rotary Microtome Low Profile Blades
Catalog # 77435-360
undefined
Rotary Microtome Low Profile Blades
Catalog # 77435-360
Supplier Number:  RFBLADES

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Rotary Microtome low profile blades
  • Size:
    80×8 mm
  • Thickness:
    0.24 mm
  • Cat. No.:
    77435-360

Specifications

About this item

Rotary microtome low-profile cutting blades for tissue sectioning.

  • Box set contains 50 individual blades in a dispenser box
  • Blade angle: 35 degrees

These low profile blades are compatable with any rotary microtome model.

Packaging: Set includes one blade dispenser box with 50 blades.