Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

SD Pro™ Primate Probiotic, Fiber Plus Tablet

SD Pro™ primate probiotic, fiber plus tablets, cherry flavor, 6 g tablet, 2.54 cm (1"), 0.64 cm (¼"), 100/bottle
Catalog # 77391-952
Supplier:   Bio-Serv
 :  PB120-100
SD Pro™ Primate Probiotic, Fiber Plus Tablet
Catalog # 77391-952
Supplier:  Bio-Serv
SD Pro™ Primate Probiotic, Fiber Plus Tablet

SD Pro™ primate probiotic, fiber plus tablets, cherry flavor, 6 g tablet, 2.54 cm (1"), 0.64 cm (¼"), 100/bottle

Price
$284.24
Log in for availability
Pack of 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

SD Pro™ is a series of species derived origin probiotics designed specifically for the host species that combines the benefits of Lactobacillus reuteri and Saccharomyces boulardii.

  • SD Pro™ tablets contains live culture, which is sensitive to heat and moisture
  • Highly palatable, sweet cherry flavored tablets

L. reuteri is a natural bacterial inhabitant of a healthy GI tract and contains antimicrobial properties. S. boulardii regulates intestinal microbial homeostasis and prevents the colonization of pathogens within the GI tract.

Each tablet contains ~1.25 billion CFU of Lactobacillus reuteri, ~140 mg Saccharomyces boulardii and 300 mg L-Glutaminein a highly palatable, easy to dose flavored tablet.

Fiber Plus includes the added benefits of 1 gm of soluble fiber (Inulin) and 1 gm of insoluble fiber (Cellulose) to aid in gastrointestinal support.

Ingredients: Sucrose, dextrose, cellulose, inulin, L-glutamine, flavoring, yeast culture, hydrolyzed yeast, calcium silicate, magnesium stearate, dried Lactobacillus reuteri, fermentation product, soybean oil, sodium aluminosilicate

Note: If non-human primate is on antibiotic therapy, administer SD Pro 4 to 6 hours after antibiotic dosage.

Specifications
  • Ø:
    2.54 cm (1")
  • Description:
    SD Pro™ primate probiotic, fiber plus tablets, cherry flavor, 6 g tablet
  • Height:
    0.64 cm (¼")
  • Packaging:
    100/bottle