Synthware Vacuum Trap Assembly with #15 O-Ring Joint, Kemtech America
Catalog # 60004-656
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Vacuum Trap Assembly, Complete
- Cat. No.:60004-656
- Supplier no.:V391557
Specifications
About this item
Designed for use with manifolds that have a #15 O-ring joint.
- Heavy wall Borosilicate construction
- 35/20 Spherical joint
The seal between upper and lower parts is made with a 57 mm flat O-ring flange. The trap body has an O.D. of 57 mm, and is 300 mm in length. Complete trap assembly is supplied with horseshoe clamp, one #331 Viton® O-ring, and two #116 Viton® O-rings. All accessory items are sold separately.