Orders LinkContactUsLinkComponent
Synthware Vacuum Traps with Spherical Joints
Synthware Vacuum Traps with Spherical Joints
Synthware Vacuum Traps with Spherical Joints
Synthware Vacuum Traps with Spherical Joints
Traps feature 35/20 spherical joints for added flexibility.
  • Heavy Wall Borosilicate Construction
  • 35/20 Spherical Joint
  • Adapters sold Separately
The trap body has a 50 mm ext. Ø and is 360 mm in length. Each of the three adapters features a 16 mm ext. Ø outlet. The three adapters and 35/20 stainless steel pinch clamps must be ordered separately.
Product Family Options
  • Description
  • Middle/pump adapter
    Each
    Each - $45.39
    $45.39
    Each
       $45.39
     : V175038
  • Top adapter
    Each
    Each - $49.89
    $49.89
    Each
       $49.89
     : V175037
  • Vacuum trap body
    Each
    Each - $196.22
    $196.22
    Each
       $196.22
     : V175036
Accessories
  •  
    Description
  • 60005-114
    Replacement 2 mm PTFE Plug
    Each - $23.02
  •  
    Description
  • 60005-116
    PTFE plug, 4 mm
    Each - $38.54
  •  
    Description
  • 60004-978
    Clamps for 35/20 spherical joints
    Each - $35.50