ContactUsLinkComponent
Synthware Flow Control Adapters with Metering Stopcock, Kemtech America
Synthware Flow Control Adapters with Metering Stopcock, Kemtech America
Catalog # 60000-306
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Flow Control Adapters with Metering Stopcock, Kemtech America
Catalog # 60000-306
Supplier:  Kemtech America
Supplier Number:  A732440
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • [ST] joint no.:
    24/40
  • Cat. No.:
    60000-306
  • Supplier no.:
    A732440
Specifications
About this item
Adapter has a metering stopcock between top and bottom [ST] joints to permit flow control of fluids between apparatus.
  • Metering stopcock
  • Top outer [ST] joint
  • Bottom inner [ST] joint
Adapter features a 2 mm PTFE metering stopcock between top outer and lower inner [ST] joints. Stopcock bore is 2 mm.