ContactUsLinkComponent
PurSwab® Chemical Resistant Foam-Over-Cotton Tipped Applicator, Puritan Medical Products
PurSwab® Chemical Resistant Foam-Over-Cotton Tipped Applicator, Puritan Medical Products
  10806-144
Product available on GSA Advantage®
undefined
PurSwab® Chemical Resistant Foam-Over-Cotton Tipped Applicator, Puritan Medical Products
  10806-144
 :  1806-WCFCR
Product available on GSA Advantage®
  • Description:
    Wood handle
  • Cat. No.:
    10806-144
  • Handle length:
    149.2 mm (5.875")
  • Head width:
    9.5 mm (0.375")
  • Head length:
    20.6 mm (0.812")
  • Supplier no.:
    1806-WCF CR
This specialty foam tipped applicator is produced with chemical resistant foam.
Foam over cotton provides extra absorbency. It is slightly textured for light abrasion and will not swell or break apart with most solvents. For use in critical environments as a general cleaning swab, removing contamination and adhesives, and applying adhesives or other solutions.
 : CE marked.
 : Packaged as 20 bags of 50 swabs per bag.