Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
SD Pro™ Mini Primate Probiotic, Magic Tabs

SD Pro™ Mini Primate Probiotic, Magic Tabs

SD Pro Mini primate probiotic, magic tabs, pina colada flavor, 2 g tablet, 1.9 cm (0.75"), 0.64 cm (¼"), 100/bottle
Catalog # 77391-956
Supplier:   Bio-Serv
Supplier #:  PB135-100
SD Pro™ Mini Primate Probiotic, Magic Tabs
Catalog # 77391-956
Supplier:  Bio-Serv
SD Pro™ Mini Primate Probiotic, Magic Tabs

SD Pro Mini primate probiotic, magic tabs, pina colada flavor, 2 g tablet, 1.9 cm (0.75"), 0.64 cm (¼"), 100/bottle

Price
$453.52
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

SD Pro magic tabs are a therapetutic cocktail of beneficial probiotics, potassium gluconate and bismuth subsalicylate for use in primates with idiopathic diarrhea. Coined 'Magic Tabs' as they are an all-in-one tasty tablet conferring similar benefits as bananas, Pepto Bismol and microbiome support.

  • Highly palatable, sweet pina colada flavored tablets
  • Shelf life: 6 months from date of manufacture - refrigerated
  • SD Pro tablets contains live culture, which is sensitive to heat and moisture

SD Pro™ is a series of species derived origin probiotics designed specifically for the host species that combines the benefits of Lactobacillus reuteri and Saccharomyces boulardii. S. boulardii regulates intestinal microbial homeostasis and prevents the colonization of pathogens within the GI tract. L. reuteri is a natural bacterial inhabitant of a healthy GI tract and contains antimicrobial properties. Each tablet contains ~0.4 billion CFU of Lactobacillus reuteri , 42.5 mg Saccharomyces boulardii, 0.65 mEq K+, 36 mg Bismuth Subsalicylate and 75 mg L-Glutamine in a highly palatable, easy to dose flavored tablet.

Ingredients: Sucrose, dextrose, L-glutamine, berry flavoring, renal K, L-glutamine, yeast culture, hydrolyzed yeast, calcium silicate, magnesium stearate, dried lactobacillus reuteri, fermentation product, soybean oil, bismuth subsalicylate, sodium aluminosilicate.

Feeding guidelines: One tablet treats an average 400 gm marmoset. Note: If non-human primate is on antibiotic therapy, administer SD Pro 4 to 6 hours after antibiotic dosage.

Specifications
  • Ø:
    1.9 cm (0.75")
  • Description:
    SD Pro Mini primate probiotic, magic tabs, pina colada flavor, 2 g tablet
  • Height:
    0.64 cm (¼")
  • Packaging:
    100/bottle
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.