Pyramid Feeder, Certified
Pyramid feeder, certified
Catalog # 76406-472
Supplier: Bio-Serv
: K1100
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Made of durable pure virgin nylon.
- Certified (contaminant screened)
- Use with any powdered, liquid, soft or gelled diet
- Provides easy access to critical care diets for sick or debiliated animals
- Autoclavable and cagewasher safe
Specifications
- Description:Pyramid feeder, certified