SD Pro™ Primate Probiotic, Powder
76353-782
: Bio-Serv
- Description:SD Pro, primate probiotic, powder
- Environmentally Preferable:
- Cat. No.:76353-782
- Packaging:75 doses/jar
SD Pro is a series of species derived origin probiotics designed specifically for the host species that combines the benefits of Lactobacillus reuteri and Saccharomyces boulardii.
- Simply top dress biscuits or mix with semi-soft food ingredients
- Shelf life: 12 months from date of manufacture, refrigerated
L. reuteri is a natural bacterial inhabitant of a healthy GI tract and contains antimicrobial propertie. S. boulardii egulates intestinal microbial homeostasis and prevents the colonization of pathogens within the GI tract.
Each scoop (4 g) contains ~2 billion CFU of Lactobacillus reuteri, ~2 billion CFU of Saccharomyces boulardii and 300 mg L-Glutamine.
Ingrediants: Yeast Extract, Yeast Culture, Dextrose, L-Glutamine, Active Dried Yeast (dried Saccharomyces cerevisiae fermentation product), Dried Lactobacillus reuteri fermentation product, Sodium Aluminosilicate
: This product is not meant to diagnose, treat, cure or prevent disease.