Synthware Addition Funnel with High Vacuum Valve, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Funnels are equipped with a high vacuum valve that features a PTFE O-ring at the tip and a PTFE wiper to protect the Viton® O-ring on the shaft.
- Heavy wall borosilicate construction
- Ground glass joint
- High vacuum valves included
Designed for the slow and controlled addition of reagents to a reaction.