Order Entry
Puerto Rico
ContactUsLinkComponent
 
Desiccating Cabinet, Boekel Scientific
  24981-008
 :  Boekel
undefined
Desiccating Cabinet, Boekel Scientific
  24981-008
 :  Boekel
 :  1344

 

  • Int. Dimensions:
    48.3W×34.3D cm (19×131/2") shelves, 30.5W×41.6D×5H cm (12×163/8×2") tray
  • Cat. No.:
    24981-008
  • Ext. dimensions:
    41.9W x 58.4L x 56.5H cm (161/2 x 23 x 221/4")
  • Shipping weight:
    27.2 kg (60 lbs.)
  • Supplier no.:
    1344

 

 

Cabinet has two height-adjustable, stainless steel shelves and a stainless steel tray for holding desiccant or instruments.

  • “Sealtight” door
  • Completely airtight and moisture-proof
  • Manual relief valve normalizes pressure in cabinet before opening door
  • Indentations on top surface for stacking and storing
  • Made of highly polished 18–8 stainless steel, with three sides of 4.75mm (3/16") glass
  • 1.4 kg (3 lbs.) tray