Polyseed® B.O.D.5 Seed Inoculum
Supplier: InterLab
Certificates
About this item
BOD Seed Inoculum for use in Biochemical Oxygen Demand (BOD5) test procedures. A blend of specialized microbial cultures in an easy-to-use capsule. Designed to provide a uniform standard for the degradation of both industrial and municipal waste in BOD5 analyses.
- Consitent seed source: same bacteria, same type of bacteria, same amount of bacteria blend
- Known bacterial content is free from pathogens
- Save and convenient pre-measured dry product in easy to use capsules
Polyseed®-NX CBOD contains an additive for the inhibition of ammonia nitrogen in a wastewater sample. Additive amount is comparable to that stated in Standard Methods as necessary for sufficient inhibition.
Two years shelf life
Packaging: Package of 50 capsules.
Related Information
Certificates
Search by Lot Number66130-430 (Datasheet)
Datasheet
66130-430 (Procedure)