Synthware Filtration Apparatus with 47 mm Stainless Steel Disc, [ST]40/35 Joint Connections, Kemtech America
Catalog # 60002-276
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Complete Support Assembly
- Material:Stainless steel
- Cat. No.:60002-276
- Supplier no.:F10401LA
Specifications
About this item
Uses 47 mm membranes for filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks.
- Complete Kit
- Heavy wall borosilicate construction
- Ground glass joint
- Stainless steel filter
Support base has removable 47 mm stainless steel support screen for use with viscous or proteinaceous solutions.
Delivery: Complete assembly includes a 300 ml graduated funnel, aluminum clamp, support base, and 1 L filtering flask.