Synthware Distilling Receivers, Kemtech America
Catalog # 60002-072
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Cat. No.:60002-072
- [ST] joint size:14/20
- Supplier no.:D381412
Specifications
About this item
With 135° angle between top outer joint and lower inner joints.
- Heavy wall Borosilicate construction
- Ground glass joint
- Keck clips included
- graduated receiver tubes included
Supplied with distribution adapter, three 12 ml receiver tubes, and three metal clamps.