ContactUsLinkComponent
Synthware Reflux Condensers, Large Cooling Capacity with Removable Hose Connections, Kemtech America
Synthware Reflux Condensers, Large Cooling Capacity with Removable Hose Connections, Kemtech America
  60000-990
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Reflux Condensers, Large Cooling Capacity with Removable Hose Connections, Kemtech America
  60000-990
 :  C534125
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
  • Height:
    300 mm
  • Cat. No.:
    60000-990
  • Coil length:
    125 mm
  • Supplier no.:
    C534125
Coil ext. Ø.
  • Nylon caps and hose connection included
  • Heavy wall borosilicate construction
  • Ground glass joint
This is 8 mm to provide maximum cooling capacity. Jacket ext. Ø: 40 mm. [ST] joints size: 24/40. Hose connection: 8 mm.