ContactUsLinkComponent
Sol-M® Luer Adapter
Sol-M® Luer Adapter
Catalog # 77528-890
undefined
Sol-M® Luer Adapter
Catalog # 77528-890
Supplier Number:  LA21920
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Luer adapter, 20G
  • Cat. No.:
    77528-890
Specifications
About this item
The Sol-M® luer adapter is designed to work in a variety of blood collection applications by securely connecting to female Luer connections.
  • Blue cap to differentiate from other blood collection devices
  • Designed to ensure secure connections
An ideal accessory to use with blood collection sets that feature a holder for evacuated tubes. Fitted with rubber sleeve to ensure no blood spills or exposure.