ContactUsLinkComponent
Synthware Chain Clamp, Kemtech America
Synthware Chain Clamp, Kemtech America
Catalog # 60005-040
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Chain Clamp, Kemtech America
Catalog # 60005-040
Supplier:  Kemtech America
Supplier Number:  W150150
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Description:
    Synthware Chain Clamp
  • Cat. No.:
    60005-040
  • Supplier no.:
    W150150
Specifications
About this item
Clamp includes chain and spring
  • 80 to150 mm clamping diameter
  • Chrome plated
Accommodates objects with a diameter from 80 to 150mm.