Order Entry
Puerto Rico
ContactUsLinkComponent
Reagent Bottles with Stoppers, Polypropylene, Wide Mouth, BrandTech
Reagent Bottles with Stoppers, Polypropylene, Wide Mouth, BrandTech
Catalog # 83008-546
Supplier:  Brandtech
undefined
Reagent Bottles with Stoppers, Polypropylene, Wide Mouth, BrandTech
Catalog # 83008-546
Supplier:  Brandtech
Supplier Number:  101794

Specifications

  • [ST] joint no.:
    45/40
  • Capacity:
    500 ml (16.9 oz.)
  • Cat. No.:
    83008-546
  • Diameter ×H:
    87×183 mm
  • Supplier no.:
    V101794

Specifications

About this item

Polypropylene reagent bottles with polypropylene stoppers provide excellent chemical resistance to many aliphatic alcohols, aldehydes, aliphatic hydrocarbons, alkalis, and acids.

  • Break-resistant
  • Polypropylene bottle and stopper for high chemical resistance
  • High translucency
  • Rugged design

Bottles and stoppers are autoclavable.