ContactUsLinkComponent
VWR UniTranz-RT® Transport System, Media Only
  77200-790
 :  
undefined
VWR UniTranz-RT® Transport System, Media Only
  77200-790
 :  
  • Media Format:
    Liquid
  • Description:
    Universal transport media
  • Size:
    3 ml
  • Cat. No.:
    77200-790
The UniTranz-RT® universal transport is ideal for the collection and preservation of viruses including, chlamydia, mycoplasma, and urea plasma. It is also suitable for use with antigen detection, PCR, and rapid tests when validated by test manufacturers and/or laboratory.
  • 3 ml fill
  • Medium is stable at room temperature
  • No swab, media only
  • Premium medical grade plastic components improve product shelf life
  • Shelf life: 18 months
  • For In Vitro diagnostic use
  • Made in USA
Contains antimicrobial agents to minimize bacterial and fungal contamination, as well as glass beads to release and disperse sample into the medium during vortexing.