TRIO.BAS™ AIRBIO ONE RAPID-VIRUS Sampling System
Catalog # 76636-296
Supplier: Hardy Diagnostics
Specifications
- Flow rate:100 L/min
- Description:AIRBIO ONE RAPID-VIRUS system
- Cat. No.:76636-296
Specifications
About this item
The AIRBIO ONE RAPID-VIRUS System is an air sampler for airborne viable particle sampling. It is designed for total pathogen surveillance of bacteria, fungi, yeast, and viruses.
- Collectionon unit is completely sterilizable
- Tri-clamp stainless steel tube system facilitates cleaning and sterilization
Liquid sampling permits rapid methods (e.g. PCR), while traditional air sampling using culture media agar plates requires incubation of media plates to obtain sample results.
The dual-purpose AIRBIO ONE RAPID-VIRUS instrument may be used to collect samples in liquid for subsequent rapid analytical identification steps by PCR, or sample air via traditional impact-to-agar culture media plate method to count the sample colonies (CFU).