TRIO.BAS™ Base Station Induction Battery Charger
Catalog # 75928-106
Supplier: Hardy Diagnostics
Specifications
- Description:Base station induction battery charger
- Cat. No.:75928-106
- Supplier no.:BAS318
Specifications
About this item
Induction battery charging base station for use with the TRIO.BAS™ MONO, DUO, TRIO, and Isolator air sampling units.
- 100/240 VCA
- 50/60 Hz
- 35 W US variant plug
Units simply rest on top of the base station and begin charging.