Vee Gee Filter Holder Systems
Supplier: Heathrow Scientific
Certificates
About this item
Filtration filter holders are offered with a wide variety of components for different applications.
- Available in 100 or 300 ml capacities
Available with a fritted glass support base for general and biological applications, or with a removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.