ContactUsLinkComponent
Accessories for Synthware Vacuum Adapters with Glass Stopcock
Catalog # 60005-162
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Accessories for Synthware Vacuum Adapters with Glass Stopcock
Catalog # 60005-162
Supplier:  Kemtech America
Supplier Number:  S461230
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Description:
    Replacement 2 mm glass plug
  • Cat. No.:
    60005-162
  • Supplier no.:
    S461230
Specifications
About this item
Kemtech all glass replacement straight bore stopcock.
  • Borosilicate 3.3
  • Straight bore
  • 12/30 plug size
  • 1:10 taper
To be used with our constant pressure retention kit.