Specifications
- Description:Jar
- Size:85 g (3 oz.)
- Cat. No.:95042-716
- Supplier no.:3-740
Specifications
About this item
Powdered formula removes stains and tarnish, and restores luster to surgical instruments, autoclaves, ultrasonic cleaners, stainless steel countertops, tiles, and plated surfaces
Effective on a variety of materials including stainless steel, chrome, nickel, brass, copper, and silver. The formula will not harm instrument surfaces.