Polytox® Rapid Biological Toxicity Screening Test, International Laboratory
Catalog # 66130-436
Supplier: Avantor
Specifications
- Cap included:Yes
- Description:Rapid Biological Toxicity Screening Test
- Cat. No.:66130-436
- Supplier no.:P-410
Specifications
About this item
Polytox® is a blend of specialized bacterial cultures designed to provide a simple, rapid, and economical test for measuring toxicity of wastewater to biological wastewater treatment systems in less than 30 minutes, without the use of expensive instrumentation.
EPA recommended. Three to six month shelf life. (InterLab P-410)