Order Entry
Puerto Rico
ContactUsLinkComponent
Somso® Pelvis with Uterus in Ninth Month of Pregnancy
Somso® Pelvis with Uterus in Ninth Month of Pregnancy
Catalog # 470347-532
Supplier:  MARCUS SOMMER
Somso® Pelvis with Uterus in Ninth Month of Pregnancy
Catalog # 470347-532
Supplier:  MARCUS SOMMER
Supplier Number:  MS13

Specifications

  • Description:
    9th month pelvis with uterus
  • Height:
    41 cm
  • Length:
    40 cm
  • Cat. No.:
    470347-532
  • Width:
    28 cm

Specifications

About this item

Somso® premium quality models.

  • Five year warranty

Natural size, in SOMSO-Plast®. The model shows the right half of the female pelvis in median section. Fetus removable. After Prof. Dr. Petry. Comprises two parts.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.