ContactUsLinkComponent
Test Tube Racks, Aluminum, Black Machine Company
Test Tube Racks, Aluminum, Black Machine Company
  60943-160
 :  Avantor
undefined
Test Tube Racks, Aluminum, Black Machine Company
  60943-160
 :  Avantor
  • No. of Holes:
    24
  • Cat. No.:
    60943-160
  • For tube O.D.:
    26 mm
  • Supplier no.:
    A100/B
Punched from hard sheet aluminum, these S-shaped racks are practical and rust-proof
The low profile and wide, flat base prevent tipping. Racks also slide into each other for compact storage. Edges and holes have smooth finish.
 : Manufactured in the USA. Custom racks are available. Contact your Avantor sales representative for more information.