Serum Vials, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Ideally suited for handling, containing, and storing a variety of liquids, including reagents, vaccines, and chromatography samples.
- Autoclavable
These crimp-top serum vials are lighter in weight than molded bottles. More closely held tolerances and uniformity of glass allow the vials to be lyophilized.
Ordering information: Vials available in clear or amber glass, without closure. Stoppers and seals sold separately.