ContactUsLinkComponent
Pre-Cleaned Straight-Sided Jars, Polyethylene, Environmental Express
Pre-Cleaned Straight-Sided Jars, Polyethylene, Environmental Express
Catalog # 23050-048
Small Business Enterprise
Pre-Cleaned Straight-Sided Jars, Polyethylene, Environmental Express
Catalog # 23050-048
Supplier Number:  BPC3240
Small Business Enterprise
Specifications
  • Cap Size:
    89-400
  • Capacity:
    1 L (33.8 oz.)
  • Description:
    Level III Jar
  • Cat. No.:
    23050-048
  • Supplier no.:
    BPC3240
Specifications
About this item
Natural high-density poylethylene jars include a polypropylene cap and F-217 PE liner.
All containers meet one of the three levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level II indicates that each case of containers has been custody-sealed and lot number-labeled for traceability to the enclosed Statement of Compliance.