Kahnetics® Deluxe Needle Sample Kit, Cooper Tools
Catalog # 37695-332
Supplier: Cooper Tools
Specifications
- Description:Kahnetics® Deluxe Needle Sample Kit
- Environmentally Preferable:
- Cat. No.:37695-332
- Supplier no.:KDS660
Specifications
About this item
Kit contains most available needle sizes and needle types, tip caps, and scabboards
Needles range in size from 14 to 30 gauge and 1.27cm (1/2") to 3.81cm (11/2") in length. Kit includes plastic and rubber tip caps, as well as Luer-lock tip caps in black (UV-blocking) and clear. Kit also includes scabboards for 14, 16, 18, 20, and 22 gauge needles. 500 piece assortment.