Orders LinkContactUsLinkComponent
PYREX® Graham Condensers, Corning
PYREX® Graham Condensers, Corning
   Sustainable Options Available
Supplier:   Corning
PYREX® Graham Condensers, Corning
Supplier:  Corning
   Sustainable Options Available
Certificates
PYREX® Graham Condensers, Corning
4 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These Graham condensers have a high surface area per unit with a standard taper outer joint at the top and a standard taper inner joint at the bottom with a drip tip. They are primarily used in vacuum distillations. Condenser tubulations are approximately 10 mm ext.Ø.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 89091-370
  • 89091-434
  • 23139-041
  • 89090-724
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental Certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Specifications
Product Family Options
Product Information
  • [ST] joint
    Jacket length
    Availability
    Price
  • 24/40
    400 mm (15¼")
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,880.00
    $1,880.00
    Case of 4
       $1,880.00
    Supplier Number: 2560-400
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-370
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental Certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • 24/40
    500 mm (19¹¹⁄₁₆")
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $2,050.00
    $2,050.00
    Case of 4
       $2,050.00
    Supplier Number: 2560-500
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-434
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental Certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 19/38
    200 mm (7⅞")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $276.00
    $276.00
    Case of 1
       $276.00
    Supplier Number: 2560-200
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    23139-041
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental Certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 24/40
    300 mm (11⁴⁄₅")
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $402.00
    $402.00
    Log in for availability
    Case of 6
    Case of 6 - $2,017.63
    $2,017.63
    Pack of 1
       $402.00
    Case of 6
       $2,017.63
    Supplier Number: 2560-300
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-724
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental Certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact