Order Entry
Puerto Rico
ContactUsLinkComponent
Synthware Adapter, Vacuum, with Stopcock, Microscale, Kemtech America
Synthware Adapter, Vacuum, with Stopcock, Microscale, Kemtech America
  60004-102
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Adapter, Vacuum, with Stopcock, Microscale, Kemtech America
  60004-102
 :  A381410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

 

  • [ST] joint no.:
    14/10
  • Cat. No.:
    60004-102
  • Tubing connection:
    8 mm
  • Bore size:
    2 mm
  • Supplier no.:
    A381410

 

 

Vacuum adapters are used to regulate the flow of gases or liquids into reation systems.

  • With stopcock
  • For microscale applications

Vacuum joint adapter features a 1:5 PTFE stopcock.