Order Entry
Puerto Rico
ContactUsLinkComponent
PureYield™ RNA Midiprep Systems, Promega
PureYield™ RNA Midiprep Systems, Promega
Certificates
undefined
PureYield™ RNA Midiprep Systems, Promega
Certificates

Specifications

  • Storage Conditions:
    15 - 30 °C
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Midi
  • Refine target molecule:
    Total RNA
  • Sample type:
    Blood
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    PureYield RNA Midiprep System

Specifications