DuPont™ Tychem® 2000 Sleeves
Catalog # 13450-338
Supplier: DuPont
Specifications
- Garment application:Food,Industrial,Laboratory,Pharmaceutical
- Gender:
- Color:Yellow
- Length:18¾"
- Environmentally Preferable:
- Cat. No.:13450-338
- Supplier no.:QC500BYL00020000
Specifications
About this item
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
- Elastic at both ends
- Bound seams
- One size fits most
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing.
These sleeves feature elastic at both bicep, measuring 81/4", and wrist, measuring 61/2".
Certifications: Thickness meets ASTM D1777.