ContactUsLinkComponent
Synthware Reflux Condensers, Kemtech America
Synthware Reflux Condensers, Kemtech America
  60000-996
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Reflux Condensers, Kemtech America
  60000-996
 :  C351922
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
  • [ST] joint:
    19/22
  • Cat. No.:
    60000-996
  • Supplier no.:
    C351922
Coil section has a height of 100 mm.
  • Heavy wall borosilicate construction
  • Ground glass joint
Coiled circulation tube for increased surface area.