Order Entry
Puerto Rico
ContactUsLinkComponent
Synthware Condenser, Air Reflux, Microscale, Kemtech America
Synthware Condenser, Air Reflux, Microscale, Kemtech America
Catalog # 60004-122
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Condenser, Air Reflux, Microscale, Kemtech America
Catalog # 60004-122
Supplier:  Kemtech America
Supplier Number:  C221420
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • [ST] joint:
    14/10
  • Length:
    143 mm
  • Cat. No.:
    60004-122
  • Supplier no.:
    C221420

Specifications

About this item

Air reflux condenser for microscale applications.

  • Heavy wall borosilicate construction
  • Ground glass joint

Length between ST joints: 96 mm.