KLEENGUARD® A20 Breathable Particle Protection Shoe Cover, Kimberly-Clark Professional®
Catalog # 10843-667
Supplier: Ansell Healthcare
Specifications
- Shoe size:XL/XXL
- Garment application:Chemical protection
- Color:Blue
- Environmentally Preferable:Waste Reducing
- Cat. No.:10843-667
- Supplier no.:66857
Specifications
About this item
Shoe cover is constructed of a unique breathable MICROFORCE™ barrier fabric for protection from particulates
These A20 breathable particle protection shoe cover are made of polyethylene.
Seamless. Nonconductive. Free of natural rubber latex.
Ordering information: Cleanroom packaged with inner liner. Bulk packed 100 pair per bag, 3 bags per case.