Human TNFRSF8 (Tumor Necrosis Factor Receptor Superfamily, Member 8) ELISA Kit
Catalog # 77528-768
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:TNFRSF8 (Tumor Necrosis Factor Receptor Superfamily, Member 8)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.32 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, CD and adhesion molecule, apoptosis, tumor immunity, reproductive science
- Sensitivity:0.135 ng/ml
- Regulatory Status:RUO
- Cat. No.:77528-768