Rat CREB (Cyclic AMP Response Element Binding Protein) ELISA Kit
Catalog # 77528-492
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:CREB (Cyclic AMP Response Element Binding Protein)
- Size:96 wells
- Sample Type:Tissue homogenates and other biological fluids.
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.32 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Epigenetics and nuclear signaling, immunology, neuroscience
- Sensitivity:0.132 ng/ml
- Regulatory Status:RUO
- Cat. No.:77528-492