Mouse PGD2 (Prostaglandin D2) ELISA Kit
77526-656
:
- Assay Type (ELISA with LOV):Competitive
- Host:Mouse
- Target protein:PGD2 (Prostaglandin D2)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Cell proliferation
- Sensitivity:4.34 pg/ml
- Regulatory Status:RUO
- Cat. No.:77526-656