Pig BAFF/CD257 (B-Cell Activating Factor) ELISA Kit
Catalog # 77525-792
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Pig
- Target protein:BAFF/CD257 (B-Cell Activating Factor)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:CD and adhesion molecule, tumor immunity, infection immunity, immune molecule
- Sensitivity:0.055 ng/ml
- Regulatory Status:RUO
- Cat. No.:77525-792