Mouse LAMa5 (Laminin Alpha 5) ELISA Kit
77524-136
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:LAMa5 (Laminin Alpha 5)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:78.13 - 5000 pg/ml
- Storage Temperature:2…8 °C
- Application:Developmental science
- Sensitivity:33 pg/ml
- Regulatory Status:RUO
- Cat. No.:77524-136