Rat CR2 (Complement Receptor 2) ELISA Kit
Catalog # 77518-112
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:CR2 (Complement Receptor 2)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids.
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:1.57 - 100 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, CD and adhesion molecule, infection immunity, immune molecule
- Sensitivity:0.57 ng/ml
- Regulatory Status:RUO
- Cat. No.:77518-112