Rat TF (Tissue Factor) ELISA Kit
Catalog # 77514-560
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:TF (Tissue Factor)
- Size:96 wells
- Sample Type:Plasma, tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:31.25 - 2000 pg/ml
- Storage Temperature:2…8 °C
- Application:Hematology, cardiovascular biology
- Sensitivity:12.4 pg/ml
- Regulatory Status:RUO
- Cat. No.:77514-560