Mouse BMP1 (Bone Morphogenetic Protein 1) ELISA Kit
Catalog # 77513-252
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:BMP1 (Bone Morphogenetic Protein 1)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:78.13 - 5000 pg/ml
- Storage Temperature:2…8 °C
- Application:Developmental science
- Sensitivity:30 pg/ml
- Regulatory Status:RUO
- Cat. No.:77513-252