Mouse CX3CL1 (Chemokine C-X3-C-Motif Ligand 1) ELISA Kit
Catalog # 77511-488
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:CX3CL1 (Chemokine C-X3-C-Motif Ligand 1)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Cytokine, infection immunity, rheumatology, autoimmunity
- Sensitivity:0.056 ng/ml
- Regulatory Status:RUO
- Cat. No.:77511-488