Rat DCN (Decorin) ELISA Kit
77510-996
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:DCN (Decorin)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:7.82 - 500 pg/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, developmental science, bone metabolism
- Sensitivity:3.1 pg/ml
- Regulatory Status:RUO
- Cat. No.:77510-996