Grade 50 Cheesecloth, Lymtech®, FG Cleanwipes
Catalog # 82001-040
Supplier: Avantor
Specifications
- Cat. No.:82001-040
- Supplier no.:GRADE 50 CC
Specifications
About this item
100% pure cotton cheesecloth is ideal for wiping, buffing, polishing, staining, and filtering.
- 28x24 thread count
Very absorbent and low linting.